3-Bromo-1,8-naphthalic anhydride 95%

Supplier: Ambeed
Warning

A634879-1G A634879-250MG A634879-5G
76727-188EA 60 USD
76727-188 76727-190 77880-344
3-Bromo-1,8-naphthalic anhydride 95%
3-Bromo-1,8-naphthalic anhydride

Precaución: for Research Use Only

Formula: C₁₂H₅BrO₃
MDL Number: MFCD00228162
CAS Number: 24050-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR