2-Chlorothioxanthone 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one

A634029-100G A634029-25G
76727-174EA 56.23 USD
76727-174 76727-176
2-Chlorothioxanthone 98%
2-Chlorothioxanthone

Precaución: for Research Use Only

Formula: C₁₃H₇ClOS
MW: 246.72 g/mol
MDL Number: MFCD00005067
CAS Number: 86-39-5
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR