Silver chloride 99%

Supplier: Ambeed
Danger

A633569-25G A633569-5G A633569-100G A633569-10G
76727-170EA 58 USD
76727-170 76727-172 77669-718 77873-029
Silver chloride 99%
Silver chloride

Precaución: for Research Use Only

Formula: AgCl
MW: 143.32 g/mol
Boiling Pt: 1554
Melting Pt: 455
MDL Number: MFCD00003399
CAS Number: 7783-90-6
UN: 3077
ADR: 9 Ⅲ
Merck Index: 13,08582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR