(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine 95%

Supplier: Ambeed
Danger

Synonyms: Paroxol

A632262-100G A632262-10G A632262-25G A632262-1G A632262-5G
76727-164EA 160.2 USD
76727-164 76727-166 76727-168 77602-594 77602-596
(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine 95%
(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine

Precaución: for Research Use Only

Formula: C₁₃H₁₈FNO
MW: 223.29 g/mol
MDL Number: MFCD06658161
CAS Number: 105812-81-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR