4-Methyl-1-piperazinecarbonyl chloride hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: 4-Methyl-1-piperazinecarbonyl Chloride Hydrochloride

A630574-10G A630574-25G A630574-5G
76727-148EA 9.55 USD
76727-148 76727-150 76727-152
4-Methyl-1-piperazinecarbonyl chloride hydrochloride 95%
4-Methyl-1-piperazinecarbonyl chloride hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClN₂O·HCl
MW: 199.08 g/mol
MDL Number: MFCD01074875
CAS Number: 55112-42-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR