Sodium mesitylenesulfonate 98%
Supplier: Ambeed
|
Warning
|
A629169-1G
A629169-25G
A629169-5G
76727-132EA
13
USD
76727-132
76727-134
76727-136
Sodium mesitylenesulfonate 98%
Sodium mesitylenesulfonate
Precaución: for Research Use Only
Formula:
C₉H₁₂O₃S |
MDL Number:
MFCD00067712 CAS Number: 6148-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...