Potassium-2-(methylthio)phenyltrifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(2-(methylthio)phenyl)borate, Potassium(2-Methylsulphonylphenyl)trifluoroboroate

A629056-1G A629056-5G
76727-128EA 13 USD
76727-128 76727-130
Potassium-2-(methylthio)phenyltrifluoroborate 98%
Potassium 2-(methylthio)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KS
MDL Number: MFCD04115773
CAS Number: 850623-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR