Phenyl 1-thio-β-D-galactopyranoside 97%

Supplier: Ambeed
Warning

Synonyms: Phenyl 1-thio-β-D-galactoside, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol

A628547-100MG A628547-250MG A628547-5G A628547-1G
76727-120EA 15.92 USD
76727-120 76727-122 76727-124 77602-588
Phenyl 1-thio-β-D-galactopyranoside 97%
Phenyl 1-thio-β-D-galactopyranoside

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₅S
MW: 272.317 g/mol
MDL Number: MFCD00063272
CAS Number: 16758-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR