Sodium dihydrogen phosphate 99%

Supplier: Ambeed
Warning

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic

A627079-1000G A627079-100G A627079-500G
76727-100EA 17 USD
76727-100 76727-102 76727-104
Sodium dihydrogen phosphate 99%
Sodium dihydrogen phosphate

Precaución: for Research Use Only

Formula: NaH₂PO₄
MW: 119.98 g/mol
Melting Pt: 650
MDL Number: MFCD00003527
CAS Number: 7558-80-7
Merck Index: 12,08806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR