Nickel (II) sulfamate tetrahydrate ≥98%

Supplier: Ambeed
Danger

Synonyms: Nickel (II) sulphamidate tetrahydrate, Nickel bis(sulphamidate) tetrahydrate, Nickel sulphamate tetrahydrate, Nickel(II)sulphamate tetrahydrate

A626175-25G A626175-100G
76727-098EA 5.31 USD
76727-098 77602-582
Nickel (II) sulfamate tetrahydrate ≥98%
Nickel(II) sulphamate tetrahydrate

Precaución: for Research Use Only

Formula: H₄N₂NiO₆S₂·4H₂O
MW: 322.92 g/mol
MDL Number: MFCD00149966
CAS Number: 124594-15-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR