N-Benzylcinchonidinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: (2S,4S,5R)-1-Benzyl-2-((R)-hydroxy(quinolin-4-yl)methyl)-5-vinylquinuclidin-1-ium chloride, BCDC

A623205-10G A623205-1G A623205-25G A623205-5G
76727-082EA 98.67 USD
76727-082 76727-084 76727-086 76727-088
N-Benzylcinchonidinium chloride 98%
N-Benzylcinchonidinium chloride

Precaución: for Research Use Only

Formula: C₂₆H₂₉ClN₂O
MW: 420.98 g/mol
MDL Number: MFCD00082422
CAS Number: 69257-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR