N-(2-Cyanoethyl)-N-butylaniline 98+%

Supplier: Ambeed
Warning

Synonyms: N-Butyl-N-(2-cyanoethyl)aniline, 3-(Butyl(phenyl)amino)propanenitrile, 3-(N-Butyl-N-phenylamino)propionitrile

A622154-100G A622154-25G A622154-5G
76727-076EA 143 USD
76727-076 76727-078 76727-080
N-(2-Cyanoethyl)-N-butylaniline 98+%
N-(2-Cyanoethyl)-N-butylaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂
MDL Number: MFCD00019863
CAS Number: 61852-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR