N,N-Diethylcyclohexylamine 98%

Supplier: Ambeed
Danger

A620952-1G A620952-5G A620952-25G
76727-072EA 9.55 USD
76727-072 76727-074 77669-672
N,N-Diethylcyclohexylamine 98%
N,N-Diethylcyclohexylamine

Precaución: for Research Use Only

Formula: C₁₀H₂₁N
MW: 155.2804 g/mol
MDL Number: MFCD00003847
CAS Number: 91-65-6
UN: 2734
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR