Potassium tris(1-pyrazolyl)borohydride hydrate 98%

Supplier: Ambeed
Warning

A619532-250MG A619532-5G A619532-1G A619532-25G
76727-056EA 18 USD
76727-056 76727-058 77569-178 77880-324
Potassium tris(1-pyrazolyl)borohydride hydrate 98%
Potassium tris(1-pyrazolyl)borohydride hydrate

Precaución: For research use only.

Formula: C₉H₁₀BKN₆·xH₂O
MDL Number: MFCD03414149
CAS Number: 18583-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR