N-(4-Hydroxyphenyl)maleimide 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Hydroxyphenyl)-1H-pyrrole-2,5-dione

A617669-1G A617669-5G A617669-25G A617669-10G A617669-100G
76727-048EA 5 USD
76727-048 76727-050 77602-558 77669-660 77880-320
N-(4-Hydroxyphenyl)maleimide 98%
N-(4-Hydroxyphenyl)maleimide

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MDL Number: MFCD00129167
CAS Number: 7300-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR