N-Methyltaurine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylamino)ethanesulphonic acid

A617506-100MG A617506-10G A617506-1G A617506-250MG A617506-25G A617506-5G A617506-100G
76727-036EA 6.37 USD
76727-036 76727-038 76727-040 76727-042 76727-044 76727-046 77551-852
N-Methyltaurine 95%
N-Methyltaurine

Precaución: for Research Use Only

Formula: C₃H₉NO₃S
MW: 139.1735 g/mol
MDL Number: MFCD00014735
CAS Number: 107-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR