Sodium formate 98%

Supplier: Ambeed
Warning

A617155-1000G A617155-100G A617155-500G
76727-030EA 21 USD
76727-030 76727-032 76727-034
Sodium formate 98%
Sodium formate

Precaución: for Research Use Only

Formula: HCO₂Na
MDL Number: MFCD00013101
CAS Number: 141-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR