Sodium methanesulfonate 97%

Supplier: Ambeed
Warning

A616866-100G A616866-10G A616866-500G A616866-25G A616866-1000G
76727-024EA 17 USD
76727-024 76727-026 76727-028 76844-368 77669-650
Sodium methanesulfonate 97%
Sodium methanesulphonate

Precaución: for Research Use Only

Formula: CH₃NaO₃S
MDL Number: MFCD00064389
CAS Number: 2386-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR