2-Aminoethyl dihydrogen phosphate 98%

Supplier: Ambeed
Danger

Synonyms: O-Phosphorylethanolamine

A615303-25G A615303-5G
76727-022EA 23 USD
76727-022 77573-004
2-Aminoethyl dihydrogen phosphate 98%
2-Aminoethyl dihydrogen phosphate

Precaución: for Research Use Only

Formula: C₂H₈NO₄P
MW: 141.0630 g/mol
MDL Number: MFCD00008178
CAS Number: 1071-23-4
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR