Aluminium potassium sulfate dodecahydrate 98%

Supplier: Ambeed
Warning

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

A614658-1000G A614658-100G A614658-500G
76727-016EA 11.67 USD
76727-016 76727-018 76727-020
Aluminium potassium sulfate dodecahydrate 98%
Aluminium potassium sulphate dodecahydrate

Precaución: for Research Use Only

Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
MDL Number: MFCD00149143
CAS Number: 7784-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR