1-Benzyl-4-hydroxypiperidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Benzylpiperidin-4-ol
A612978-100G
A612978-10G
A612978-25G
A612978-500G
A612978-5G
A612978-1000G
76727-000EA
32
USD
76727-000
76727-002
76727-004
76727-006
76727-008
77669-634
1-Benzyl-4-hydroxypiperidine 98%
1-Benzyl-4-hydroxypiperidine
Precaución: for Research Use Only
Formula:
C₁₂H₁₇NO MW: 191.27 g/mol |
MDL Number:
MFCD00006503 CAS Number: 4727-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...