Procaine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Diethylamino)ethyl 4-Aminobenzoate, 4-Aminobenzoic acid 2-(Diethylamino)ethyl ester, 2-(Diethylamino)ethyl 4-aminobenzoate

A610960-100G A610960-10G A610960-25G A610960-500G A610960-5G
76726-978EA 56.23 USD
76726-978 76726-980 76726-982 76726-984 76726-986
Procaine 98%
Procaine

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₂
MW: 236.31 g/mol
MDL Number: MFCD00007893
CAS Number: 59-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR