N-isopropyl-2-pyrrolidone 98%

Supplier: Ambeed
Danger

A609754-1G A609754-250MG A609754-5G
76726-968EA 53 USD
76726-968 76726-970 76726-972
N-isopropyl-2-pyrrolidone 98%
N-isopropyl-2-pyrrolidone

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MDL Number: MFCD00015398
CAS Number: 3772-26-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR