Sodium hexafluorophosphate 99%

Supplier: Ambeed
Danger

A608607-25G A608607-5G
76726-964EA 37 USD
76726-964 76726-966
Sodium hexafluorophosphate 99%
Sodium hexafluorophosphate

Precaución: for Research Use Only

Formula: F₆NaP
MDL Number: MFCD00011122
CAS Number: 21324-39-0
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR