N-(tert-Butyl)-2-chloroacetamide 97%

Supplier: Ambeed
Warning

A606727-1G A606727-25G A606727-5G
76726-956EA 14.86 USD
76726-956 76726-958 76726-960
N-(tert-Butyl)-2-chloroacetamide 97%
N-(tert-Butyl)-2-chloroacetamide

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
MDL Number: MFCD00032565
CAS Number: 15678-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR