N-acetyl-4-S-cysteaminylphenol 98%

Supplier: Ambeed
Warning

A605768-100MG A605768-1G A605768-250MG A605768-5G
76726-938EA 27 USD
76726-938 76726-940 76726-942 76726-944
N-acetyl-4-S-cysteaminylphenol 98%
N-acetyl-4-S-cysteaminylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂S
MDL Number: MFCD07779474
CAS Number: 91281-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR