p-Tolyl trifluoromethanesulfonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Methylphenyl Trifluoromethanesulphonate, Trifluoromethanesulphonic acid p-Tolyl Ester, p-Tolyl Triflate
A605446-1G
A605446-25G
A605446-5G
76726-932EA
15.92
USD
76726-932
76726-934
76726-936
p-Tolyl trifluoromethanesulfonate 98%
p-Tolyl trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₈H₇F₃O₃S MW: 240.20 g/mol |
MDL Number:
MFCD00192507 CAS Number: 29540-83-8 UN: 2922 ADR: 8,6.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...