2,9-Di(tridecan-7-yl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 95%

Supplier: Ambeed
Warning

A603765-1G A603765-250MG
76726-924EA 267 USD
76726-924 76726-926
2,9-Di(tridecan-7-yl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 95%
2,9-Di(tridecan-7-yl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone

Precaución: for Research Use Only

Formula: C₅₀H₆₂N₂O₄
MDL Number: MFCD08702668
CAS Number: 110590-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR