3'-Methylpropiophenone 98%

Supplier: Ambeed
Warning

A600794-100G A600794-1G A600794-25G A600794-5G
76726-908EA 519.85 USD
76726-908 76726-910 76726-912 76726-914
3'-Methylpropiophenone 98%
3'-Methylpropiophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2017 g/mol
MDL Number: MFCD00027014
CAS Number: 51772-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR