3-(1-Naphthyl)propionic acid 96%

Supplier: Ambeed
Warning

A597211-250MG A597211-25G A597211-1G A597211-5G
76726-904EA 18 USD
76726-904 76726-906 77572-924 77669-578
3-(1-Naphthyl)propionic acid 96%
3-(1-Naphthyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
MDL Number: MFCD00033058
CAS Number: 3243-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR