Potassium sorbate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate
A594220-100G
A594220-500G
A594220-25G
A594220-1000G
76726-866EA
6
USD
76726-866
76726-868
77572-912
77669-568
Potassium sorbate 95%
Potassium sorbate
Precaución: for Research Use Only
Formula:
CH₃CH=CHCH=CHCOOK MW: 150.22 g/mol Melting Pt: 270 |
MDL Number:
MFCD00016546 CAS Number: 24634-61-5 Merck Index: 13,07756 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...