Monobutyl phthalate 95%

Supplier: Ambeed
Warning

A593783-10G A593783-1G A593783-25G A593783-5G
76726-858EA 107 USD
76726-858 76726-860 76726-862 76726-864
Monobutyl phthalate 95%
Monobutyl phthalate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MDL Number: MFCD00043498
CAS Number: 131-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR