2-Methoxy-N,N-dimethylacetamide 97%

Supplier: Ambeed
Warning

A592608-1G A592608-5G A592608-100G A592608-25G
76726-854EA 11 USD
76726-854 76726-856 77669-558 77669-560
2-Methoxy-N,N-dimethylacetamide 97%
2-Methoxy-N,N-dimethylacetamide

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MW: 117.14 g/mol
MDL Number: MFCD07787542
CAS Number: 4128-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR