Sodium tetrafluoroborate 99%

Supplier: Ambeed
Danger

A591292-25G A591292-100G
76726-834EA 17 USD
76726-834 77594-800
Sodium tetrafluoroborate 99%
Sodium tetrafluoroborate

Precaución: for Research Use Only

Formula: BF₄Na
MDL Number: MFCD00003515
CAS Number: 13755-29-8
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR