Methyltributylammonium chloride 97%

Supplier: Ambeed
Warning

Synonyms: Tributylmethylammonium chloride, N,N-Dibutyl-N-methylbutan-1-aminium chloride

A591129-100G A591129-10G A591129-25G A591129-500G A591129-1000G
76726-826EA 20 USD
76726-826 76726-828 76726-830 76726-832 77602-468
Methyltributylammonium chloride 97%
Methyltributylammonium chloride

Precaución: for Research Use Only

Formula: C₁₃H₃₀ClN
MW: 235.84 g/mol
MDL Number: MFCD00011847
CAS Number: 56375-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR