N-Methyl-M-toluidine 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Methylamino)toluene, N,3-Dimethylaniline

A588447-25G A588447-5G
76726-812EA 13 USD
76726-812 76726-814
N-Methyl-M-toluidine 98%
N-Methyl-M-toluidine

Precaución: for Research Use Only

Formula: C₈H₁₁N
MW: 121.18 g/mol
MDL Number: MFCD00035787
CAS Number: 696-44-6
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR