(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid 98%

Supplier: Ambeed
Warning

A587591-10MG A587591-1G A587591-1MG A587591-50MG
76726-788EA 20 USD
76726-788 76726-790 76726-792 76726-794
(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid 98%
(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₄
MDL Number: MFCD00069231
CAS Number: 28860-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR