Potassium (4-(cbz-amino)phenyl)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium 4-(benzyloxycarbonylamino)phenyltrifluoroborate

A586969-1G A586969-250MG A586969-5G
76726-780EA 53.05 USD
76726-780 76726-782 76726-784
Potassium (4-(cbz-amino)phenyl)trifluoroborate 98%
Potassium (4-(Cbz-amino)phenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₁₄H₁₂BF₃KNO₂
MW: 333.16 g/mol
MDL Number: MFCD04115743
CAS Number: 850623-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR