(2S,3aS,6aS)-1-(((S)-1-Carboxy-3-phenylpropyl)-L-alanyl)octahydrocyclopenta[b]pyrrole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Ramiprilat

A586852-10MG A586852-1MG A586852-25MG
76726-774EA 186.72 USD
76726-774 76726-776 76726-778
(2S,3aS,6aS)-1-(((S)-1-Carboxy-3-phenylpropyl)-L-alanyl)octahydrocyclopenta[b]pyrrole-2-carboxylic acid 95%
(2S,3aS,6aS)-1-(((S)-1-Carboxy-3-phenylpropyl)-L-alanyl)octahydrocyclopenta[b]pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₈N₂O₅
MW: 388.46 g/mol
MDL Number: MFCD00865787
CAS Number: 87269-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR