2-(4-Fluorophenyl)indole 98%

Supplier: Ambeed
Warning

A585984-250MG A585984-1G
76726-758EA 8 USD
76726-758 77872-965
2-(4-Fluorophenyl)indole 98%
2-(4-Fluorophenyl)indole

Precaución: for Research Use Only

Formula: C₁₄H₁₀FN
MDL Number: MFCD00068178
CAS Number: 782-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR