Octafluoro-4,4'-biphenol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Octafluorobiphenyl-4,4'-diol, Octafluoro-4,4'-dihydroxybiphenyl
A583498-1G
A583498-250MG
A583498-25G
A583498-5G
76726-748EA
59.42
USD
76726-748
76726-750
76726-752
76726-754
Octafluoro-4,4'-biphenol 95%
Octafluoro-4,4'-biphenol
Precaución: for Research Use Only
Formula:
C₁₂H₂F₈O₂ MW: 330.13 g/mol |
MDL Number:
MFCD00045743 CAS Number: 2200-70-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...