(S)-Ethyl 2-hydroxy-4-phenylbutanoate ≥98%

Supplier: Ambeed
Warning

A582007-100MG A582007-1G A582007-250MG A582007-5G
76726-736EA 18.04 USD
76726-736 76726-738 76726-740 77669-530
(S)-Ethyl 2-hydroxy-4-phenylbutanoate ≥98%
(S)-Ethyl 2-hydroxy-4-phenylbutanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.25 g/mol
MDL Number: MFCD03095465
CAS Number: 125639-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR