3-(4-Nitrophenoxy)benzoic acid 98%

Supplier: Ambeed
Warning

A579715-1G A579715-250MG A579715-5G
76726-724EA 34 USD
76726-724 76726-726 76726-728
3-(4-Nitrophenoxy)benzoic acid 98%
3-(4-Nitrophenoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₉NO₅
MW: 259.22 g/mol
MDL Number: MFCD00690134
CAS Number: 27237-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR