Pentaerythritol tetraacrylate 80% (by GC) stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A577035-1000G A577035-100G A577035-500G A577035-25G
76726-712EA 123 USD
76726-712 76726-714 76726-716 77551-734
Pentaerythritol tetraacrylate 80% (by GC) stabilized
Pentaerythritol tetraacrylate

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₈
MDL Number: MFCD00015334
CAS Number: 4986-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR