2,3:5,6-Di-O-isopropylidene-D-mannofuranose 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(+)-2,3:5,6-Di-O-isopropylidene-α-D-mannofuranose, 2,3:5,6-Di-O-isopropylidene-D-mannofuranose, (3aS,4S,6R,6aS)-6-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-ol, D-Mannose diacetonide
A576241-100G
A576241-1G
A576241-25G
A576241-5G
76726-704EA
249.32
USD
76726-704
76726-706
76726-708
76726-710
2,3:5,6-Di-O-isopropylidene-D-mannofuranose 98%
2,3:5,6-Di-O-isopropylidene-D-mannofuranose
Precaución: for Research Use Only
Formula:
C₁₂H₂₀O₆ MW: 260.29 g/mol |
MDL Number:
MFCD00134206 CAS Number: 14131-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...