Sodium pyrene-1-sulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Pyrenesulphonic acid sodium

A574821-100MG A574821-1G A574821-250MG
76726-692EA 40.32 USD
76726-692 76726-694 76726-696
Sodium pyrene-1-sulfonate 98%
Sodium pyrene-1-sulfonate

Precaución: for Research Use Only

Formula: C₁₆H₉NaO₃S
MW: 304.29 g/mol
MDL Number: MFCD00210088
CAS Number: 59323-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR