(S)-N-(1-Amino-5-guanidino-1-oxopentan-2-yl)benzamide hydrochloride 95%

Supplier: Ambeed
Warning

A574785-1G A574785-250MG A574785-5G
76726-686EA 30.77 USD
76726-686 76726-688 76726-690
(S)-N-(1-Amino-5-guanidino-1-oxopentan-2-yl)benzamide hydrochloride 95%
(S)-N-(1-Amino-5-guanidino-1-oxopentan-2-yl)benzamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₀ClN₅O₂
MW: 313.78 g/mol
MDL Number: MFCD00035003
CAS Number: 4299-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR