Sclareol 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2R,4aS,8aS)-1-((R)-3-Hydroxy-3-methylpent-4-en-1-yl)-2,5,5,8a-tetramethyldecahydronaphthalen-2-ol

A573023-100MG A573023-1G A573023-1MG A573023-250MG A573023-25MG A573023-50MG A573023-5G A573023-25G A573023-100G
76726-670EA 14.86 USD
76726-670 76726-672 76726-674 76726-676 76726-678 76726-680 76726-682 77602-410 77669-510
Sclareol 97%
Sclareol

Precaución: for Research Use Only

Formula: C₂₀H₃₆O₂
MW: 308.51 g/mol
MDL Number: MFCD00869558
CAS Number: 515-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR