Methyl-4-(trifluoromethyl)pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(trifluoromethyl)nicotinate

A572033-100MG A572033-1G
76726-668EA 16 USD
76726-668 77594-734
Methyl-4-(trifluoromethyl)pyridine-3-carboxylate 97%
Methyl-4-(trifluoromethyl)pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
MDL Number: MFCD00101568
CAS Number: 175204-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR