(4-Benzyl-1,4-oxazinan-2-yl)methylamine 98%

Supplier: Ambeed
Danger

Synonyms: (4-Benzylmorpholin-2-yl)methylamine

A571542-1G A571542-250MG A571542-5G
76726-656EA 59 USD
76726-656 76726-658 76726-660
(4-Benzyl-1,4-oxazinan-2-yl)methylamine 98%
(4-Benzyl-1,4-oxazinan-2-yl)methylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O
MW: 206.29 g/mol
MDL Number: MFCD02682010
CAS Number: 110859-47-7
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR